Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Placental lactogen Monoclonal Antibody (INN-hPL-37)
GREENER_CHOICE

Catalog No. PIMA125113
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125113 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125113 Supplier Invitrogen™ Supplier No. MA125113
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-25113 detects Human placental lactogen (epitope 2) from human samples.

The human growth hormone (GH) locus is comprised by two GH (GH1 and GH2) genes and three chorionic somatomammotropin (CSH1, CSH2 and CSH-L) genes. Placental lactogens (CSH1, CSH2) are produced in the mammalian placenta and are similar in structure and function to growth hormones. Together, placental lactogens and growth factors play an essential role to assure successful lactation after pregnancy. Placental lactogens also modify the metabolic state of the mother during pregnancy to supply energy to the fetus. Both CSH1 and CSH2 can be detected by this antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Placental lactogen
Applications Immunoradiometric Assay
Classification Monoclonal
Clone INN-hPL-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.09% sodium azide
Gene CSH1
Gene Accession No. P0DML2, P0DML3
Gene Alias Choriomammotropin; chorionic somatomammotropin A; chorionic somatomammotropin hormone 1; chorionic somatomammotropin hormone 1 (placental lactogen); Chorionic somatomammotropin hormone 2; chorionic somatomammotropin-1; CS-1; CSA; CSH1; CSH2; CSMT; GHB3; growth hormone B3; hCS-1; hCS-A; hPL; Lactogen; PL; placental lactogen
Gene Symbols CSH1, CSH2
Host Species Mouse
Immunogen Human placental lactogen (hPL).
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1442, 1443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.