Learn More
CSH1 Mouse anti-Human, Clone: OTI2E8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506791

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the somatotropin/prolactin family of hormones and plays an important role in growth control. The gene is located at the growth hormone locus on chromosome 17 along with four other related genes in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. Although the five genes share a remarkably high degree of sequence identity, they are expressed selectively in different tissues. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed mainly in the placenta and utilizes multiple transcription initiation sites. Expression of the identical mature proteins for chorionic somatomammotropin hormones 1 and 2 is upregulated during development, although the ratio of 1 to 2 increases by term. Mutations in this gene result in placental lactogen deficiency and Silver-Russell syndrome.Specifications
CSH1 | |
Monoclonal | |
Unconjugated | |
CSH1 | |
CS-1, CSA, CSMT, GHB3, PL, hCS-1, hCS-A | |
Mouse | |
Affinity Chromatography | |
RUO | |
1442 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2E8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P0DML2 | |
CSH1 | |
Full length human recombit protein of human CSH1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.