Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597634
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597634 Supplier Invitrogen™ Supplier No. PA597634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSHL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

May be a novel gestational hormone required to compensate for absence of other members of the GH/CS cluster during gestation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSHL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CSHL1
Gene Accession No. Q14406
Gene Alias chorionic somatomammotropin CS-5; chorionic somatomammotropin hormone like 1; chorionic somatomammotropin hormone-like 1; Chorionic somatomammotropin-like; CS-5; CSHL1; CSHP1; CSL; GHB4; growth hormone B4; growth hormone cluster; hCS-L; Lactogen-like
Gene Symbols CSHL1
Host Species Rabbit
Immunogen Recombinant Human Chorionic somatomammotropin hormone-like 1 protein (1-128AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1444
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.