Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSK Antibody (CSK-04), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500344F
Description
CSK Monoclonal antibody specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
CSK | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry, Immunoprecipitation | |
CSK-04 | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450. | |
0.1 mL | |
Cell Cycle and Replication, Signal Transduction | |
1445 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction