Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSK Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100808
Description
CSK Polyclonal specifically detects CSK in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
CSK | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Reported variants represent identical protein: NP_004374.1; NP_001120662.1. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_004374.1 | |
CSK | |
Peptide with sequence C-EQLEHIKTHELHL corresponding to C-Terminus according to NP_004374.1, NP_001120662.1. | |
0.1 mg | |
Cell Cycle and Replication, Signal Transduction | |
1445 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction