Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSK Antibody (SR1874), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32226850UL
Description
CSK Monoclonal antibody specifically detects CSK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CSK | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1874 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
A synthesized peptide derived from human CSK (Uniprot #: P41240) | |
50 μg | |
Cell Cycle and Replication, Signal Transduction | |
1445 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction