Learn More
CSK Binding Protein Antibody (PAG-C1) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP142418
Catalog No. NBP142418
Only null left
Add to Cart
Description
Description
CSK Binding Protein Monoclonal antibody specifically detects CSK Binding Protein in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).
Specifications
Specifications
CSK Binding Protein
Monoclonal
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
C-terminal peptide (last 15 amino acids) of human Csk binding protein coupled to KLH.
0.05 mg
Primary
Human, Mouse, Rat, Bovine
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
PAG-C1
Unconjugated
Mouse
Protein A or G purified
RUO
The antibody recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp), also known as protein associated with glycosphingolipid-enriched microdomains (PAG).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only