Learn More
Promega CSK Kinase Enzyme System, 1mg
Catalog No. PRV2982 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2982 1 mg
1 options
Catalog No. PRV2982 Supplier Promega Supplier No. V2982
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CSK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CSK has been shown to downregulate the tyrosine kinase activity of c-src through tyrosine phosphorylation of the c-src carboxy terminus.
  • Easily Screen and Profile CSK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C