Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSK Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298358R
Description
CSK Polyclonal specifically detects CSK in Mouse samples. It is validated for Western Blot, ELISA.Specifications
CSK | |
Polyclonal | |
Western Blot, ELISA | |
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
Produced in rabbits immunized with purified, recombinant CSK (P41241; Met1-Leu450). | |
0.1 mL | |
Cell Cycle and Replication, Signal Transduction | |
1445 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction