Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
csl/RBPJK Rabbit anti-Human, Mouse, Clone: 9B3L6, Novus Biologicals™

Rabbit Monoclonal Antibody
$196.25 - $452.15
Specifications
| Antigen | csl/RBPJK |
|---|---|
| Clone | 9B3L6 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
csl/RBPJK Monoclonal antibody specifically detects csl/RBPJK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| csl/RBPJK | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CBF-1, csl, H-2K binding factor-2, IGKJRB1RBPSUHCBF1, IGKJRBRBP-J kappa, immunoglobulin kappa J region recombination signal binding protein 1, J kappa-recombination signal-binding protein, KBF2, RBP-JK, RBPJKMGC61669, RBP-Jrecombining binding protein suppressor of hairless (Drosophila), recombination signal binding protein for immunoglobulin kappa J region, recombining binding protein suppressor of hairless, Renal carcinoma antigen NY-REN-30, SUH, suppressor of hairless homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human csl/RBPJK (Q06330). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9B3L6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience, Signal Transduction, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3516 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title