Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-His-Leu-Lys-Trp-Ura

Catalog No. mp114550022
Click to view available options
Quantity:
100 g

CSM-His-Leu-Lys-Trp-Ura, Powder

  • 5 dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.
  • Time tested formulation is available in complete and dropout formulations
  • For use with DOB or YNB with the appropriate nitrogen and carbon sources to custom formulate your own selective Synthetic Defined yeast media
  • 5 Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified
Content And Storage Store CSM-HIS-LEU-LYS-TRP-URA powder at ambient temperature (15-30°C). CSM-HIS-LEU-LYS-TRP-URA powder is hygroscopic, therefore minimize exposure to air to prolong storage of powder mixture. Store sterile agar medium at 4°C and liquid medium at 15-30°C in subdued light.
Description 5 dropout of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiaegrowth media. Time tested formulation is available in complete and dropout formulations.
Form Powder
Product Type Cell Culture Media
Sterility Sterile
Formulation Drop-Out Supplementsmg/L Adenine 0 L-Arginine HCl 50 L-Aspartic Acid 80 L-Histidine HCl 0 L-Isoleucine 50 L-Leucine 0 L-Lysine HCl 50 L-Methionine 20 L-Phenylalanine 50 L-Threonine 100 L-Tryptophan 0 L-Tyrosine 50 Uracil 0 L-Valine 140
For Use With (Application) Used for S. cerevisiae growth media
pH 4.0 to 4.4
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.