Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSMD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552587
Description
Immunogen sequence: RYSAPYCSLP RAPLHGFILG QTSTQPGGSI HFGCNAGYRL VGHSMAICTR HPQGYHLWSE AIPLCQALSC GLPEAPKNGM VFGKEYTVGT KAMYSCSEGY HLQAGAEATA ECLDTGLWSN RNVPPQCVPV TCPDVSSISV EH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Specifications
CSMD2 | |
Polyclonal | |
Unconjugated | |
CSMD2 | |
B230311I21; B230378I17; CSMD2; CUB and Sushi (SCR repeat) domain; CUB and sushi domain-containing protein 2; CUB and Sushi multiple domains 2; CUB and sushi multiple domains protein 2; dJ1007G16.1; dJ1007G16.2; dJ947L8.1; Gm139; KIAA1884; mKIAA1884; RGD1565728 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z408 | |
CSMD2 | |
Recombinant protein corresponding to Human CSMD2. Recombinant protein control fragment (Product #RP-90164). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction