Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSMD2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8187R

Specifications
CSMD2 | |
Polyclonal | |
Unconjugated | |
CSMD2 | |
CSMD2; CUB and Sushi (SCR repeat) domain; CUB and sushi domain-containing protein 2; CUB and Sushi multiple domains 2; CUB and sushi multiple domains protein 2; dJ1007G16.1; dJ1007G16.2; dJ947L8.1; KIAA1884 | |
Rabbit | |
Protein A | |
RUO | |
114784 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q7Z408 | |
CSMD2 | |
KLH conjugated synthetic peptide derived from human CSMD2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction