Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSMD3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093387 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093387 Supplier Novus Biologicals Supplier No. NB10093387
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CSMD3 Polyclonal specifically detects CSMD3 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen CSMD3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_937756.1
Gene Alias CUB and sushi domain-containing protein 3, CUB and Sushi multiple domains 3, KIAA1894CUB and sushi multiple domains protein 3
Gene Symbols CSMD3
Host Species Goat
Immunogen Peptide with sequence KQRTAPKTQYTGC corresponding to internal region according to NP_937756.1, NP_937757.1, NP_443132.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114788
Test Specificity This antibody is expected to recognize all reported isoforms according to NP_937756.1; NP_937757.1 and NP_443132.2.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.