Learn More
Invitrogen™ CSMD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109598
Description
Immunogen sequence: ANSVNTASWD FPVPICRAED ACGGTMRGSS GIISSPSFPN EYHNNADCTW TIVAEPGDTI SLIFTDFQME EKYDYLEIEG SEPPTIWLSG MNIPPPIISN KNWLRLHFVT DSNHRYRGFS APYQGSSTLT HTTSTGELEE HNRTTTGAIA VASTPADVTV SSVTAVTIHR LSEEQRVQVT SLRNSGLDPN TSKDGLSPHP ADTQSTRRRP RHAEQIERTK E.
CUB and Sushi multiple domains 3 (CSMD3) is a gene that encodes a protein with an unknown function. Missense mutations, silent mutations, nonsense mutations, and frameshift deletions are observed in cancers such as lung cancer, skin cancer, and stomach cancer.
Specifications
CSMD3 | |
Polyclonal | |
Unconjugated | |
CSMD3 | |
4930500N14Rik; CSMD3; CUB and sushi domain-containing protein 3; CUB and Sushi multiple domains 3; CUB and sushi multiple domains protein 3; Kiaa1894; mKIAA1894 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
114788 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2.94 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7Z407 | |
CSMD3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 220-440 of human CSMD3 (NP_937756.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.