Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556035 Supplier Invitrogen™ Supplier No. PA556035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPANGLRYGD DYVVGQNVSY MCQPGYTMEL NGSRIRTCTI NGTWSGVMPT CRAVTCPTPP QISNGRLEGT NFDWGFSISY ICSPGYELSF PAVLTCVGN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 49%.

CUB and Sushi multiple domains 3 (CSMD3) is a gene that encodes a protein with an unknown function. Missense mutations, silent mutations, nonsense mutations, and frameshift deletions are observed in cancers such as lung cancer, skin cancer, and stomach cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSMD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSMD3
Gene Accession No. Q7Z407
Gene Alias 4930500N14Rik; CSMD3; CUB and sushi domain-containing protein 3; CUB and Sushi multiple domains 3; CUB and sushi multiple domains protein 3; Kiaa1894; mKIAA1894
Gene Symbols CSMD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CSMD3. Recombinant protein control fragment (Product #RP-95004).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.