Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ csn Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144518
Description
csn Polyclonal Antibody for Western Blot, ELISA
Aids in the defense against invading fungal pathogens by degrading their cell wall chitosan.
Specifications
csn | |
Polyclonal | |
Unconjugated | |
Chitosanase; csn; RBAU_3106 | |
Recombinant Bacillus velezensis UCMB5033 Chitosanase protein (37-278aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1.161 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction