Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSN1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100362APC
Description
CSN1 Polyclonal antibody specifically detects CSN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CSN1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
COP9 signalosome complex subunit 1, COPS1JAB1-containing signalosome subunit 1, CSN1MGC71287, G protein pathway suppressor 1SGN1, GPS-1, Protein MFH, Signalosome subunit 1 | |
Maps to a region between residue 400 and the C-terminus (residue 471) of human G protein Pathway Suppressor 1 using the numbering given in Swiss-Prot entry Q13098 (GeneID 2873). | |
0.1 mL | |
Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
2873 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction