Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100362
Description
CSN1 Polyclonal specifically detects CSN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CSN1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
COP9 signalosome complex subunit 1, COPS1JAB1-containing signalosome subunit 1, CSN1MGC71287, G protein pathway suppressor 1SGN1, GPS-1, Protein MFH, Signalosome subunit 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Turkey,Bovine, Dog, Horse, Guinea pig_10141, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee,Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500- 1:2000 | |
Q13098 | |
GPS1 | |
The immunogen recognized by this antibody maps to a region between residue 400 and the C-terminus (residue 471) of human G protein Pathway Suppressor 1 using the numbering given in Swiss-Prot entry Q13098 (GeneID 2873). | |
100 μL | |
Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
2873 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction