Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSN1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988841
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8841 Supplier Bioss Supplier No. BS9126R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.

Specifications

Antigen CSN1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPS1
Gene Accession No. Q13098
Gene Alias COP9 signalosome complex subunit 1; Cops1; CSN1; G protein pathway suppressor 1; GPS1; GPS-1; JAB1-containing signalosome subunit 1; MFH; MFH protein; MGC71287; Protein MFH; R75577; SGN1; signalosome subunit 1
Gene Symbols GPS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CSN1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2873
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.