Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSN1S1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145435
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145435 200 μL
FEPA5145436 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145435 Supplier Invitrogen™ Supplier No. PA5145435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASA play an important role in the capacity of milk to transport calcium phosphate. Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSN1S1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CSN1S1
Gene Accession No. P19228, P47710
Gene Alias AA959832; alpha S1-casein B; alpha-casein; Alpha-S1-casein; alpha-s1-casein precursor; Antioxidant peptide; AW212882; Bos d 8; CASA; casein alpha s1; casein alpha-S1; casein, alpha; Casoxin-D; Csn1; CSN1S1; Csna
Gene Symbols CSN1S1
Host Species Rabbit
Immunogen Recombinant Human CSN1 protein, Arg16-Trp185 (Accession #P47710).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12990, 1446
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.