Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSN3 (CASK) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557480
Description
Immunogen sequence: PAVVRPHAQI PQRQYLPNSH PPTVVRRPNL HPSFIAIPPK KIQDKIIIPT INTIATVEPT PA Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 40%.
Kappa-casein (CSN3) stabilizes micelle formation, preventing casein precipitation in milk.
Specifications
| CSN3 (CASK) | |
| Polyclonal | |
| Unconjugated | |
| CSN3 | |
| casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07498 | |
| CSN3 | |
| Recombinant protein corresponding to Human CSN3. Recombinant protein control fragment (Product #RP-96738). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction