Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSN3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501730661 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0661 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0661 Supplier Proteintech Group Inc Supplier No. 160321AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kappa-casein stabilizes micelle formation, preventing casein precipitation in milk.

Specifications

Antigen CSN3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CSN3
Gene Accession No. P07498
Gene Alias casein kappa, CASK, CSN10, CSN3, CSNK, Kappa casein, KCA
Gene Symbols CSN3
Host Species Rabbit
Immunogen CSN3 Fusion Protein Ag8901
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1448
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.