Learn More
CSN4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300013AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Pig, Orangutan
This gene encodes one of eight subunits composing COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases.Specifications
CSN4 | |
Polyclonal | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O88544, Q9BT78 | |
Cops4 | |
Between 350 and C-term | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
COPS4 | |
CSN4, COP9 signalosome complex subunit 4, JAB1-containing signalosome subunit 4, SGN4, signalosome subunit 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26891, 51138 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.