Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSN7b Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100461FR
Description
CSN7b Polyclonal antibody specifically detects CSN7b in Human samples. It is validated for Western BlotSpecifications
CSN7b | |
Polyclonal | |
Western Blot | |
Arabidopsis, homolog) subunit 7B, COP9 constitutive photomorphogenic homolog subunit 7B (Arabidopsis), COP9 signalosome complex subunit 7b, CSN7BMGC111077, FLJ12612, Signalosome subunit 7b | |
The immunogen this antibody was made to, maps to a region between residues 225-264 (C-terminus) of human constitutive photomorphogenic homolog subunit 7B using the TrEMBL entry Q9H9Q2 (GeneID 64708). | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Neurotransmission, Signal Transduction, Transcription Factors and Regulators | |
64708 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction