Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSN7b Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113385
Description
CSN7b Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Component of the COP9 signalosome complex (CSN), a complex involved in various cellular and developmental processes. The CSN complex is an essential regulator of the ubiquitin (Ubl) conjugation pathway by mediating the deneddylation of the cullin subunits of SCF-type E3 ligase complexes, leading to decrease the Ubl ligase activity of SCF-type complexes such as SCF, CSA or DDB2. The complex is also involved in phosphorylation of p53/TP53, JUN, I-kappa-B-alpha/NFKBIA, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN-dependent phosphorylation of TP53 and JUN promotes and protects degradation by the Ubl system, respectively.
Specifications
| CSN7b | |
| Polyclonal | |
| Unconjugated | |
| COPS7B | |
| Arabidopsis; COP9 (constitutive photomorphogenic) homolog, subunit 7b; COP9 (constitutive photomorphogenic) homolog, subunit 7b (Arabidopsis thaliana); COP9 complex S7b; COP9 constitutive photomorphogenic homolog subunit 7B; COP9 signalosome complex subunit 7b; COP9 signalosome subunit 7B; Cops7b; Csn7b; D1Wsu66e; E130114M23; JAB1-containing signalosome subunit 7b; SGN7b; signalosome subunit 7b | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26895, 64708 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BV13, Q9H9Q2 | |
| COPS7B | |
| Recombinant Human COP9 signalosome complex subunit 7b protein (1-157AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction