Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSN7b Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103080
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103080 100 μL
1 options

Catalog No. PIPA5103080

Supplier: Invitrogen™ PA5103080

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COPS7B.

Component of the COP9 signalosome complex (CSN), a complex involved in various cellular and developmental processes. The CSN complex is an essential regulator of the ubiquitin (Ubl) conjugation pathway by mediating the deneddylation of the cullin subunits of SCF-type E3 ligase complexes, leading to decrease the Ubl ligase activity of SCF-type complexes such as SCF, CSA or DDB2. The complex is also involved in phosphorylation of p53/TP53, JUN, I-kappa-B-alpha/NFKBIA, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN-dependent phosphorylation of TP53 and JUN promotes and protects degradation by the Ubl system, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSN7b
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COPS7B
Gene Accession No. Q8BV13, Q9H9Q2
Gene Alias Arabidopsis; COP9 (constitutive photomorphogenic) homolog, subunit 7b; COP9 (constitutive photomorphogenic) homolog, subunit 7b (Arabidopsis thaliana); COP9 complex S7b; COP9 constitutive photomorphogenic homolog subunit 7B; COP9 signalosome complex subunit 7b; COP9 signalosome subunit 7B; Cops7b; Csn7b; D1Wsu66e; E130114M23; JAB1-containing signalosome subunit 7b; SGN7b; signalosome subunit 7b
Gene Symbols COPS7B
Host Species Rabbit
Immunogen A synthesized peptide derived from human COPS7B(Accession Q9H9Q2), corresponding to amino acid residues D179-Q229.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26895, 64708
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.