Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSNK1A1L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982490
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2490 Supplier Bioss Supplier No. BS12943R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSNK1A1/CK1 is a casein kinase. These kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins including CTNNB1, PER1, and PER2. CSNK1A1 also participates in Wnt signaling. It may play a role in segregating chromosomes during mitosis, as well as, cytoskeleton diassembly which may regulate epithelial cell migration.

Specifications

Antigen CSNK1A1L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSNK1A1L
Gene Accession No. Q8N752
Gene Alias casein kinase 1 alpha 1 like; casein kinase 1, alpha 1-like; casein kinase I alpha S-like; Casein kinase I isoform alpha-like; CK1; CKI-alpha-like; CSNK1A1L
Gene Symbols CSNK1A1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CSNK1A1L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.