Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CSNK1E (CK1 epsilon) [R178C], GST Tag Recombinant Protein

Catalog No. A33498
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

CSNK1E serine/threonine kinase is a member of the CK1 family which plays an important role in diverse cell processes, including DNA replication and repair. CSNK1E is a regulator of Yes-associated protein (YAP) transcription coactivator, which is a key regulator of organ size and a candidate human oncogene. CSNK1E is activated by CCK2R and then phosphorylates PKD2 at Ser244. Phosphorylation of PKD2 leads to its nuclear accumulation and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells. CSNK1E can phosphorylate topoisomerase (topo) II alpha at serine-1106 and this regulates the enzyme activity and sensitivity to topo II-targeted drugs.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49674
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1454
Molecular Weight (g/mol) 73.6 kDa
Name Human CSNK1E (CK1 epsilon) [R178C], GST Tag
Quantity 10 μg
Gene Alias CK1
Common Name CK1 epsilon (R178C Mutant)
KinaseFamily CK1 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Mutant
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.