Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSNK1E Monoclonal Antibody (OTI2B3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA803780

Description
Casein kinases are a group of ubiquitous Ser/Thr kinases that phosphorylate key regulatory proteins involved in the control of cell differentiation, proliferation, chromosome segregation, circadian rhythms, and metabolic pathways. CK1 family members share a highly conserved kinase domain but differ in their variable N-and C-terminal domains. Mammals have seven family members: alpha, beta, gamma1, gamma2, gamma3, delta and epsilon. They all share at least 50% amino acid identity within the protein kinase catalytic domain. CK1 isoforms exclusively use ATP as phosphate donor and are generally co-factor independent. Casein kinase I epsilon is a Wnt-regulated kinase, and regulated phosphorylation of Dvl allows fine tuning of the Wnt-b-catenin signaling pathway. CKIe positively regulates Akt possibly by inhibiting PP2A. CKIe play an important role in neurodegenerative diseases.Specifications
CSNK1E | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P49674 | |
CSNK1E | |
Human recombit protein fragment corresponding to amino acids 153-416 of human CSNK1E produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2B3 | |
Unconjugated | |
CSNK1E | |
CKIepsilon, HCKIE | |
Mouse | |
Affinity Chromatography | |
RUO | |
1454 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction