Learn More
CSNK1G1 Mouse anti-Human, Clone: OTI4F12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806333

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha′ beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.Specifications
CSNK1G1 | |
Monoclonal | |
Unconjugated | |
CSNK1G1 | |
CK1gamma1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53944 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4F12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HCP0 | |
CSNK1G1 | |
Human recombit protein fragment corresponding to amino acids 163-422 of human CSNK1G1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.