Learn More
CSNK1G2 Mouse anti-Human, Clone: OTI2F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802864

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.Specifications
CSNK1G2 | |
Monoclonal | |
Unconjugated | |
CSNK1G2 | |
CK1g2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1455 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P78368 | |
CSNK1G2 | |
Human recombit protein fragment corresponding to amino acids 163-415 of human CSNK1G2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.