Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSNK2A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109601 Supplier Invitrogen™ Supplier No. PA5109601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPGPAAGSRA RVYAEVNSLR SREYWDYEAH VPSWGNQDDY QLVRKLGRGK YSEVFEAINI TNNERVVVKI LKPVKKKKIK REVKILENLR GGTNIIKLID TVKDPVSKTP ALVFEYINNT DFKQLYQILT DFDIRFYMYE LLKALDYCHS KGIMHRDVKP HNVMIDHQQK KLRLIDWGLA EFYHPAQEYN VRVASRYFKG PELLVDYQMY DYSLDMWSLG CMLASMIFRR EPFFHGQDNY DQLVRIAKVL GTEELYGYLK KYHIDLDPHF NDILGQHSRK RWENFIHSEN RHLVSPEALD LLDKLLRYDH QQRLTAKEAM EHPYFYPVVK EQSQPCADNA VLSSGLTAAR.

Casein kinase II is a protein serine/threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions. The holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit. CK2 functions as a tetrameric complex consisting of two regulatory beta-subunits and two catalytic units (alpha and alpha') in a homomeric or heteromeric conformation. Whilst the alpha- and alpha'-subunits are catalytically identical, proteins that regulate CK2, such as cdc2 and Hsp90, preferentially bind to the alpha and not the alpha'-subunit. CK2 can phosphorylate a number of key intracellular signaling proteins implicated in tumor suppression (p53 and PTEN) and tumorigenesis (myc, jun, NF-kappaB). CK2 is also thought to influence Wnt signaling via beta-catenin phosphorylation and the PI 3-K signaling pathway via the phosphorylation of Akt.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSNK2A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSNK2A2
Gene Accession No. O54833, P19784
Gene Alias 1110035J23Rik; C77789; casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 2; casein kinase 2, alpha prime polypeptide; casein kinase II subunit alpha'; casein kinase II, alpha 2, polypeptide; CK II alpha'; CK2; CK2A2; CK2alpha'; CSNK2A1; CSNK2A2
Gene Symbols CSNK2A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human CSNK2A2 (NP_001887.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13000, 1459, 307641
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.