Learn More
Invitrogen™ CSNK2A3 Polyclonal Antibody

Description
CSNK2A3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CSNK2A3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CSNK2A3 |
| Gene Accession No. | Q8NEV1 |
| Gene Alias | casein kinase 2 alpha 3; casein kinase 2, alpha 1 polypeptide pseudogene; casein kinase 2, alpha 1 polypeptide-like; casein kinase 2, alpha 3 polypeptide; Casein kinase II alpha 1 polypeptide pseudogene; Casein kinase II subunit alpha 3; CK II alpha 3; CSNK2A1P; CSNK2A3 |
| Gene Symbols | CSNK2A3 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.