Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSNK2B Mouse anti-Human, Clone:1H8A5, Abnova™

Catalog No. 89333131 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-131 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-131 Supplier Abnova Corporation Supplier No. MAB16639
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CSNK2B.

This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus. [provided by RefSeq]

Specifications

Antigen CSNK2B
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1H8A5
Conjugate Unconjugated
Description casein kinase 2, beta polypeptide
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CSNK2B
Gene Alias CK2B, CK2N, CSK2B, G5A, MGC138222, MGC138224
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-215 of human CSNK2B from E. coli.
Quantity 100 μg
Gene ID (Entrez) 1460
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.