Learn More
Invitrogen™ CSP alpha Polyclonal Antibody

Description
Positive Samples: Mouse brain, Mouse liver, Rat brain; Cellular Location: Cell membrane, Lipid-anchor, Melanosome, Membrane.
Specifications
Specifications
| Antigen | CSP alpha |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Dnajc5 |
| Gene Accession No. | P60904, P60905 |
| Gene Alias | 2610314I24Rik; AU018536; Ceroid-lipofuscinosis neuronal protein 4; CLN4; CLN4B; Csp; CSP alpha; cysteine string protein; cysteine string protein alpha; DnaJ (Hsp40) homolog, subfamily C, member 5; DnaJ heat shock protein family (Hsp40) member C5; dnaJ homolog subfamily C member 5; Dnajc5; DNAJC5A; mir-941-2; mir-941-3; mir-941-4; mir-941-5; NCL |
| Gene Symbols | Dnajc5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.