Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSP Antibody, Novus Biologicals™
SDP

Catalog No. NB10074397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074397 Supplier Novus Biologicals Supplier No. NB10074397

Rabbit Polyclonal Antibody

CSP Polyclonal specifically detects CSP in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen CSP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS containing 1mg/ml BSA with 0.05% Sodium Azide
Gene Alias CSP, Cysteine string protein, cysteine string protein alpha, DKFZp434N1429, DKFZp761N1221, DnaJ (Hsp40) homolog, subfamily C, member 5, dnaJ homolog subfamily C member 5, DNAJC5A, FLJ00118, FLJ13070
Gene Symbols DNAJC5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80331
Test Specificity Cysteine String Protein alpha
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.