Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSPG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575459 Supplier Invitrogen™ Supplier No. PA575459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Neuroglycan C is a transmembrane chondroitin sulfate proteoglycan that is exclusively expressed in the central nervous system. NGC gene expression is developmentally regulated being altered by addiction to psychostimulants and by nerve lesion. Its core protein has a particular multidomain structure differing from those of other known proteoglycans, and this protein is modified post-translationally in various ways such as phosphorylation and glycosylation. Neuroglycan C is a novel part-time proteoglycan that changes its structure from a proteoglycan form to a non-proteoglycan form without chondroitin sulfate chains during the development of the cerebellum and retina. Evidence suggests that Neuroglycan C is involved in neuronal circuit formation in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSPG5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CSPG5
Gene Accession No. O95196, Q71M36, Q9ERQ6
Gene Alias acidic leucine-rich EGF-like domain-containing brain protein; Caleb; chondroitin sulfate proteoglycan 5; chondroitin sulfate proteoglycan 5 (neuroglycan C); Cspg5; MGC44034; Neuroglycan C; Ngc
Gene Symbols CSPG5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 221-270 of Human CSPG5.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10675, 29873, 50568
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.