Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSPP Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226101
Description
CSPP Polyclonal specifically detects CSPP in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.Specifications
CSPP | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CSPP1 | |
Peptide with sequence C-RQNIDAYHNPDARTY corresponding to internal region according to NP_079066.5. | |
0.1 mg | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:128000 | |
Centrosome And Spindle Pole Associated Protein 1, Centrosome And Spindle Pole-Associated Protein 1, CSPP1, FLJ22490 | |
Goat | |
Affinity Purified | |
RUO | |
79848 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction