Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545896 Supplier Invitrogen™ Supplier No. PA545896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AEDKAELESD PPYMEMKGKL SAKLSENSKI LISMAKENIP PNSQQTRGSL Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 90%; Rabbit: 100%; Rat: 100%.

CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSPP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CSPP1
Gene Accession No. Q1MSJ5
Gene Alias 2310020J12Rik; 4930413O22Rik; AA387550; BC055915; centrosome and spindle pole associated protein 1; Centrosome and spindle pole-associated protein 1; centrosome spindle pole associated protein 1; centrosome/spindle pole-associated protein 2; centrosome/spindle pole-associated protein 3; CSPP; CSPP1; Cspp2; Cspp3; JBTS21; RGD1307055
Gene Symbols CSPP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CSPP1 (aa 53-102).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79848
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.