Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597318 Supplier Invitrogen™ Supplier No. PA597318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSPKGLDAAT FQNVHDFNEL KDRDSETRVD LKFMYLDPPR DHHTLEIQQQ ALLREQQKRL NRIKMQEGAK VDLDAIPSAK VREQRMPRDD TSDFLKNSLL ESDSAFIGAY GETYPAIEDD VLPPPSQLPS ARERRRNKWK GLDIDSSRPN VAPDGLSLKS ISSVNVDELR VRNEERMRRL NEFHNKPINT DDESSLVDPD DIMKHIGDDG SNSVATEPWL RPGTSETLKR FMAEQLNQEQ QQIPGKPGTF TWQGLSTAHG; Positive Samples: HeLa, Jurkat, K-562; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle, spindle pole.

CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSPP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSPP1
Gene Accession No. B2RX88, Q1MSJ5
Gene Alias 2310020J12Rik; 4930413O22Rik; AA387550; BC055915; centrosome and spindle pole associated protein 1; Centrosome and spindle pole-associated protein 1; centrosome spindle pole associated protein 1; centrosome/spindle pole-associated protein 2; centrosome/spindle pole-associated protein 3; CSPP; CSPP1; Cspp2; Cspp3; JBTS21; RGD1307055
Gene Symbols CSPP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 962-1221 of human CSPP1 (NP_0790665).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211660, 79848
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.