Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSPP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988905
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8905 Supplier Bioss Supplier No. BS9208R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.

Specifications

Antigen CSPP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSPP1
Gene Accession No. Q1MSJ5
Gene Alias 2310020J12Rik; 4930413O22Rik; AA387550; BC055915; BOS_13899; Centrosome and spindle pole associated protein 1; Centrosome and spindle pole-associated protein 1; centrosome spindle pole associated protein 1; centrosome/spindle pole-associated protein 2; centrosome/spindle pole-associated protein 3; CSPP; Cspp1; Cspp2; Cspp3; JBTS21; LOC100345834; RGD1307055
Gene Symbols CSPP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CSPP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79848
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.