Learn More
CSRP1 Antibody, Biotin, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP299738B
Catalog No. NB062635
Only null left
Add to Cart
Description
Description
CSRP1 Polyclonal specifically detects CSRP1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
CSRP1
Polyclonal
Western Blot, ELISA
CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein
Produced in rabbits immunized with purified, recombinant CSRP1 (NP_004069.1; Met 1-Glu 193).
0.1 mL
Primary
Human
Purified
Western Blot, ELISA
Biotin
PBS
Rabbit
Affinity purified
RUO
1465
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.