Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSRP1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298292R
Description
CSRP1 Polyclonal specifically detects CSRP1 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
CSRP1 | |
Polyclonal | |
Western Blot, ELISA | |
CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein | |
Produced in rabbits immunized with purified, recombinant CSRP1 (P97315; Met1-Glu193). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
1465 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction