Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSRP2BP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA549650

Catalog No. PA549650


Description

Description

CSRP2 is a protein containing two LIM domains, which are double zinc finger motifs found in proteins of diverse function. CSRP2 and some related proteins are thought to act as protein adapters, bridging two or more proteins to form a larger protein complex. The protein encoded by this gene binds to one of the LIM domains of CSRP2 and contains an acetyltransferase domain. Although the encoded protein has been detected in the cytoplasm, it is predomitly a nuclear protein. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CSRP2BP
Polyclonal
PBS with 0.09% sodium azide
Q9H8E8, Q8CID0
KAT14
KLH conjugated synthetic peptide between 246-275 amino acids from the Central region of human CSRP2BP.
400 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KAT14
Cysteine-rich protein 2-binding protein, CSRP2-binding protein, ADA2A-containing complex subunit 2, ATAC2, CRP2-binding partner, CRP2BP, CSRP2BP
Rabbit
Antigen affinity chromatography
RUO
228714, 57325
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.