Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109602 Supplier Invitrogen™ Supplier No. PA5109602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSRP3 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

This gene encodes a member of the CSRP family of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this protein is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Mutations in this gene are thought to cause heritable forms of hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM) in humans. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSRP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Csrp3
Gene Accession No. P50461, P50462, P50463
Gene Alias cardiac LIM domain protein; cardiac LIM protein; Clp; CMD1M; CMH12; CRP3; Csrp3; cysteine and glycine rich protein 3; cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; LMO4; MGC14488; MGC61993; MLP; MMLP; Muscle LIM protein; muscle lim protein isoform
Gene Symbols Csrp3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117505, 13009, 8048
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.