Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CST 530, Phenol
GREENER_CHOICESDP

Catalog No. 806650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 80-665-0 Supplier Tocris Bioscience™ Supplier No. 8066/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Functionalized IAP ligand for PROTACs™

CST 530, Phenol is a functionalized IAP (XIAP and cIAP) ligand for PROTAC™ research and development. Supplied with a phenol functional handle for ready conjugation to a linker/target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material tert-Butyl ((S)-1-(((S)-1-cyclohexyl-2-((2S,4S)-4-(3-hydroxyphenoxy)-2-(((R)-1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)pyrrolidin-1-yl)-2-oxoethyl)amino)-1-oxopropan-2-yl)(methyl)carbamate
CAS 2241489-43-8
Quantity 50 mg
Target Degrader Building Blocks
Molecular Formula C38H52N4O7
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.