Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CST1 Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550887
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550887 200 μL
FEMA550888 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550887 Supplier Invitrogen™ Supplier No. MA550887
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CST1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CST1
Gene Accession No. P01037
Gene Alias 1810034E21Rik; alcadein alpha; alcadein-alpha; alc-alpha; calsyntenin 1; calsyntenin-1; Clstn1; Cs1; CST1; Cst-1; Cstn1; C-terminal fragment 1-alpha; CTF1-alpha; Cystain-SA-I; cystatin 1; cystatin SA-I; cystatin SN; cystatin-1; Cystatin-SN; cysteine proteinase inhibitor, type 2 family; Kiaa0911; SAlc-alpha; Salivary cystatin-SA-1; Soluble Alc-alpha
Gene Symbols CST1
Host Species Mouse
Immunogen Recombinant Human CST1 protein, Trp21-Lys114 + VRS (Accession #P01037).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1469
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.