Learn More
Invitrogen™ CST1 Polyclonal Antibody

Description
Positive test controls include: A-431, A-549, BT-474, THP-1. The target is usually found in the following locations: Secreted. Immunogen sequence: AWSPKEEDRI IPGGIYNADL NDEWVQRALH FAISEYNKAT KDDYYRRPLR VLRARQQTVG GVNYFFDVEV GRTICTKSQP NLDTCAFHEQ PELQKKQLCS FEIYEVPWEN RRSLVKSRCQ ES.
Specifications
Specifications
| Antigen | CST1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CST1 |
| Gene Accession No. | P01037 |
| Gene Alias | 1810034E21Rik; alcadein alpha; alcadein-alpha; alc-alpha; calsyntenin 1; calsyntenin-1; Clstn1; Cs1; CST1; Cst-1; Cstn1; C-terminal fragment 1-alpha; CTF1-alpha; Cystain-SA-I; cystatin 1; cystatin SA-I; cystatin SN; cystatin-1; Cystatin-SN; cysteine proteinase inhibitor, type 2 family; Kiaa0911; SAlc-alpha; Salivary cystatin-SA-1; Soluble Alc-alpha |
| Gene Symbols | CST1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.