Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562783 Supplier Thermo Scientific Supplier No. PA562783
Add to Cart
Add to Cart

CST11 Polyclonal Antibody

CST11 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen CST11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CST11
Gene Accession No. Q9H112
Gene Alias CST11; CST8L; CTES2; cystatin 11; cystatin-11; cystatin-related epididymal-specific protein; dJ322G13.6; SC13; SC13delta
Gene Symbols CST11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CST11. Recombinant protein control fragment (Product #RP-98478).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.